Learn More
American Research Products Inc Fish Tumor Necrosis Factor a
Catalog No. 5000329779
Encompass
Fish Tumor Necrosis Factor a (TNF-a) ELISA KIT from CUSABIO Detection Range 125 pg/mL-5000 pg/mL Sensitivity 125 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29779 Supplier American Research Products Inc Supplier No. CSBE13254FH
Fish Tumor Necrosis Factor a (TNF-a) ELISA KIT from CUSABIO Detection Range 125 pg/mL-5000 pg/mL Sensitivity 125 pg/mL Sample serum plasma Method Competitive