Learn More
3M™ Probed Reusable Respirator Assembly 6100Q
Catalog No. 179873 Shop All 3M Products
Change view
Click to view available options
Quantity:
4/Cs.
Catalog No. Quantity
17-987-3 4/Cs.
1 options
Do not use for respiratory protection in contaminated area. These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Catalog No. 17-987-3 Supplier 3M™ Supplier No. 7000148501
Add to Cart
Add to Cart
Do not use for respiratory protection in contaminated area. These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
Includes
Probed facepiece and filters
Used for conducting quantitative fit tests on 6000 Series respirators.
  • Size: small
  • 3M Catalog# 6100Q
TRUSTED_SUSTAINABILITY
Specifications
Product Type Probed Respirator Assembly
For Use With (Application) Quantitative Fit Testing
Includes Probed facepiece and P100 filters
Model 6100Q
Quantity 4/Cs.