Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ 850 Homogenizer
GREENER_CHOICE

Catalog No. 15340169 Shop All Fisherbrand Products
Change view
Click to view available options
Speed:
5000 to 25,000 rpm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Speed
15-340-169 5000 to 25,000 rpm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-340-169
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Homogenizer only. Does not include stand (S1000).

The Fisher Scientific™ 850 Homogenizer is specifically designed to disrupt, emulsify or homogenize a wide range of samples in volumes ranging from 0.2mL to 10L.

  • Powerful 850w motor
  • Integrated digital speed control, displays speed and run time
  • Variable speed control from 5,000 to 25,000rpm
  • Compatible with stainless steel and disposable generator probes
  • Processing range from 0.2mL to 10L

Ordering Information:This part number (15-340-169) does not include the Generator Probes, Adapter, Tips, or Stand. They must be ordered separately.

Compliance

CE Approved

Warranty and Services

2 year warranty

Specifications

Product Type Homogenizer
Speed 5000 to 25,000 rpm
Voltage 115 V
Volume Range (Metric) 0.2 mL to 10 L
Diameter (English) 2.75 in.
Diameter (Metric) 6.9 cm
Height (English) 8.5 in.
Height (Metric) 21.5 cm
For Use With (Application) Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics
Includes Homogenizer only. Does not include Generator Probes, Adapter, Tips, or Stand. (Sold separately.)
Weight (English) 56 oz.
Weight (Metric) 1.6 kg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.