Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Bovine Serum Albumin (Lyophilized Powder)

Catalog No. BP67110 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP671-10 10 g
BP671-1 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. BP671-10
  • $928.50
    $748.65 / Each of 1

    Save $179.85

    19% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 9048-46-8

Bovine Serum Albumin is used as a protein stabilizer and blocking agent.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protein Stabilizer and Blocking Agent
Name BSA
pH Range 5 to 5.4
Quantity 10 g
Source Bovine
Packaging Amber Glass
Form Lyophilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.