Learn More
Description
Ready-to-use DNA size standards are stable at room temperature—no need to thaw and refreeze
- Specially engineered to provide blunt-end DNA of precise length for better band quality than classic markers
- Ideal for size assessment and quantitative determination of DNA mass
- Contain loading dye to reduce pipetting steps and save time
- Have two or three high intensity reference bands for better sizing
- Provided in sufficient quantity to load 100 lanes
Specifications
Specifications
| Color | Colorless |
| Form | Liquid |
| Formulation | Premixed with loading dye |
| For Use With (Application) | General purpose, large digested DNA |
| No. of Bands | 13 |
| Quantity | 100 Lanes |
| Storage Requirements | Stable at room temperature for two years. |
| Sufficient For | 100 Lanes |
| Product Type | DNA Molecular Weight Marker |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.