Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ EZ-Run™ Rec Protein Ladder

Catalog No. BP3602500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
2 x 500 μL, 200 Loadings
500 μL, 100 Loadings
Sufficient For:
100 Loadings
200 Loadings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
BP3602500 500 μL, 100 Loadings 100 Loadings
BP36021 2 x 500 μL, 200 Loadings 200 Loadings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP3602500

missing translation for 'validMainframeMsg'

Mixture of 14 highly purified recombinant proteins optimized for precise sizing of proteins by SDS-PAGE and Western blotting

  • Ladder is ideal for precise molecular weight determination of polypeptides in denaturing PAGE and Western blots
  • Each protein contains an integral Strep-tag™ II sequence1 and may be detected on Western blots using Strep-Tactin™-AP conjugate2 or an antibody against Strep-tag II
  • 50kDa protein band has greater intensity than other bands to serve as a reference
  • Supplied in gel loading buffer

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Formulation Supplied in gel-loading buffer
For Use With (Application) Molecular weight determination of polypeptides analyzed in various Gel system
Kit Contents <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet
Quantity 500 μL, 100 Loadings
Range 10 to 200 kd
Storage Requirements −20°C
Sufficient For 100 Loadings
Product Type Protein Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.