Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ EZ-Run™ Prestained Protein Marker

Catalog No. BP36011 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
2 x 500 μL, 200 Loadings
500 μL, 100 Loadings
Sufficient For:
100 Loadings
200 Loadings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
BP36011 2 x 500 μL, 200 Loadings 200 Loadings
BP3601500 500 μL, 100 Loadings 100 Loadings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP36011

missing translation for 'validMainframeMsg'

Ideal for monitoring protein separation during electrophoresis without staining and for verifying electroblotting of proteins onto membranes

  • Comprised of six purified proteins covalently coupled to a blue chromophore
  • Optimized to produce well-defined blue bands after SDS-PAGE
  • Supplied in gel loading buffer
  • 20 to 118kDa

The covalently coupled chromophore affects protein mobility; this prestained marker should only be used for approximating the molecular weight of separated proteins.

The Prestained Protein Marker is designed for monitoring protein separation during electrophoresis without staining. It is also used to verify electroblotting of proteins onto membranes.

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Formulation Supplied in gel-loading buffer
For Use With (Application) Monitoring protein separation during electrophoresis
Kit Contents <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet
Quantity 2 x 500 μL, 200 Loadings
Range 20 to 118 kd
Storage Requirements −20°C
Sufficient For 200 Loadings
Product Type Protein Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.