Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EZ-Run™ Protein Gel Solutions, Fisher BioReagents™

Catalog No. BP7710500 Shop All Fisherbrand Products
Change view
Click to view available options
Concentration:
10%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
BP7710500 10%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP7710500

missing translation for 'validMainframeMsg'

Resolution equal to or better than gradient precast gels

A ready-to-use premixed solution of acrylamide, bisacrylamide, buffer and SDS that enables ultra-fine resolution of protein bands by denaturing PAGE. This novel formulation provides the resolution of a gradient gel with less preparative work than a standard discontinuous gel system.

  • Ready-to-pour EZ-Run Protein Gel Solution for 1-D or 2-D PAGE—just add APS and TEMED
  • Wide separation range—from 3 to 200kDa on same minigel
  • Continuous gel system—no stacking gel necessary
  • Proprietary (patent-pending) gel chemistry

EZ-Run Protein Gel Solution comes with an optimized proprietary running buffer; other traditional SDS-PAGE buffers should not be used with the EZ-Run gel solution.

Applications:

Compatible with post-electrophoresis applications, such as Western blotting, MALDI analysis, protein sequencing. Suitable for all commonly used gel staining methods including Coomassie™ blue, silver staining and fluorescent dyes.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Protein Gel Solution
Composition 10% protein gel solution
Concentration 10%
Quantity 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.