Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ EZ-Run™ Protein Marker

Catalog No. BP3600500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
2 x 500 μL, 200 Loadings
500 μL, 100 Loadings
Sufficient For:
100 Loadings
200 Loadings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sufficient For
BP3600500 500 μL, 100 Loadings 100 Loadings
BP36001 2 x 500 μL, 200 Loadings 200 Loadings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP3600500

missing translation for 'validMainframeMsg'

Mixture of seven purified proteins optimized to produce uniform, compact bands when analyzed by SDS-PAGE and stained with Coomassie™ Blue

  • Supplied in gel loading buffer for direct application to SDS-PAGE gels
  • Ideal for molecular weight determination of polypeptides analyzed in various gel systems

TRUSTED_SUSTAINABILITY

Specifications

Form Liquid
Formulation Supplied in gel-loading buffer
For Use With (Application) Monitoring protein separation during SDS-PAGE
Kit Contents <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet
Quantity 500 μL, 100 Loadings
Range 14.4 to 116 kd
Storage Requirements −20°C
Sufficient For 100 Loadings
Product Type Protein Molecular Weight Marker
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.