Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ exACTGene™ dNTPs

Catalog No. BP2595200 Shop All Fisherbrand Products
Change view
Click to view available options
Concentration:
10 mM
pH:
7.3 to 7.7
Product Type:
dNTP Mix
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration pH Product Type Quantity
BP2595200 10 mM 7.3 to 7.7 dNTP Mix 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP2595200

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ultrapure dNTPs ideal for use in PCR, DNA labeling, and DNA sequencing

  • >99% triphosphate
  • Chemically synthesized to meet a high purity standard
  • Ensure consistent results and optimum sensitivity for many rigorous applications
  • Tested for purity by HPLC, performance in long-template PCR, and absence of DNase, RNase, and protease activity to ensure dependable performance and lot-to-lot consistency
  • Ultrapure dNTPs will enhance results for the following procedures: QPCR, high-fidelity PCR, long template (>8kb) PCR, RT-PCR, fluorescent sequencing, cDNA synthesis, Pyrosequencing™, and in vitro mutagenesis

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7.3 to 7.7
Product Type dNTP Mix
Content And Storage -20°C
Concentration 10 mM
Purity >99%
Quantity 200 μL
Color Colorless
For Use With (Application) PCR, DNA labeling and DNA sequencing
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.