Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Molecular Weight Markers

Catalog No. BP255150 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 μg
Size Range:
100 to 2000 bp
Product Type:
100 bp DNA Ladder
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Size Range Product Type
BP2551-50 50 μg 100 to 2000 bp 100 bp DNA Ladder
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP2551-50

missing translation for 'validMainframeMsg'

Suitable for determining the size of DNA fragments

  • Compatible with both conventional and pulsed field gel electrophoresis
  • DNase-, RNase-, and protease-free

TRUSTED_SUSTAINABILITY

Specifications

Color Colorless
Size Range 100 to 2000 bp
Form Liquid
For Use With (Application) Determining the size of DNA fragment
Quantity 50 μg
Product Type 100 bp DNA Ladder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.