Learn More
Description
- Compatible with both conventional and pulsed field gel electrophoresis
- DNase-, RNase-, and protease-free
Specifications
Specifications
| Color | Colorless |
| Size Range | 100 to 2000 bp |
| Form | Liquid |
| For Use With (Application) | Determining the size of DNA fragment |
| Quantity | 50 μg |
| Product Type | 100 bp DNA Ladder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.