Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ Nucleotides

Catalog No. BP25641 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
dNTP Set
Quantity:
100 μL
400 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
BP25641 dNTP Set 100 μL
BP25644 dNTP Set 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP25641

Nucleotides for PCR with a purity level of >98% triphosphate

  • Functionally tested in PCR
  • Can be used in dideoxy sequencing, mutagenesis, and cDNA synthesis applications
  • DNase-, RNase-, and protease-free
  • PCR dNTP Mix is a solution of dATP, dCTP, dGTP, and dTTP
  • dNTP Set contains one tube each of dATP, dCTP, dGTP, and dTTP (4 tubes/set)

TRUSTED_SUSTAINABILITY

Specifications

Format Liquid
pH 7 to 7.5
Product Type dNTP Set
Concentration 100 mM
Purity >98%
Quantity 100 μL
Color Colorless
For Use With (Application) Dideoxy sequencing, mutagenesis and cDNA synthesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.