Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ OptiGrow™ Super Broth Microbiological Media

Catalog No. BP143210B1 Shop All Fisherbrand Products
Change view
Click to view available options
Super broth; 10 x 1L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BP143210B1 Super broth; 10 x 1L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP143210B1

missing translation for 'validMainframeMsg'

Classic media in ready-to-use liquid form

  • Sterilized by autoclaving
  • For high yield of bacterial culture
  • No dusty powders to cause environmental concerns
  • Fully automatic manufacturing process eliminates unexpected contamination

  • LB Broth. Standard bacteriological nutrient medium for the propagation of E. coli for purposes of strain maintenance (sub-culturing), cloning, plasmid propagation, and protein expression
  • Super Broth. Specifically developed to increase the yield of high copy number plasmids derived from suspension cultures of E. coli strains
  • Terrific Broth. Supports higher cell densities than LB medium, increases plasmid DNA yield

Order Info

Supplied in case of ten 1L bottles

TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Form Liquid
Manufacturing Origin United States
Media Type Super broth
Fill Volume 10 x 1000 mL
Packaging Type Bottle
Quantity 10 x 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.