Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ SurePrep™ Nuclear or Cytoplasmic RNA Purification Kit

Catalog No. BP280550 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
75 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP280550 75 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP280550

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Includes

Lysis solution (20mL); Wash solution (22mL); Binding solution (25mL); RNA elution solution (6mL); Spin columns (50); Collection tubes (50); Elution tubes (1.7mL) (50); Product insert (1)

Rapid method for the isolation and purification of both nuclear and cytoplasmic RNA from cultured animal cells and animal tissue samples

  • Fast and easy processing using rapid spin-column format
  • High quality intact RNA can be used in various downstream applications
  • Enrich for cytoplasmic or nuclear RNA depending on downstream application
  • No genomic DNA contamination in the cytoplasmic fraction
  • No phenol or chloroform extractions
  • Isolate all sizes of RNA from each fraction, including all small RNA molecules (>200 nt)
  • Kit provides sufficient reagents to perform either 50 cytoplasmic RNA preparations or 25 cytoplasmic and nuclear preparations

Applications:

Purification of Nuclear and Cytoplasmic RNA suitable for real-time PCR, reverse transcription PCR, Northern blotting, RNase protection and primer extension, and expression array assays.

TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
For Use With (Application) RT-PCR, reverse transcription PCR, northern blotting, RNase protection and primer extension and expression array assays
Includes Lysis solution 20mL, wash solution 22mL, binding solution 25mL, RNA elution solution 6mL, spin columns 50, collection tubes 50, elution tubes 1.7mL 50, product insert 1 50μg Column Binding Capacity, 600μL Max. column lo
Quantity 75 Preps
Yield 35μg Grape leaves (50mg), 32μg plum leaves (50mg), 60μg tobacco leaves (50mg), 60μg tomato leaves (50mg)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.