Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ SurePrep™ RNA/DNA/Protein Purification Kit

Catalog No. BP280250 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP280250 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP280250

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Lysis solution (40mL); RNA wash solution (20mL); RNA elution solution (6mL); gDNA wash solution (15mL); gDNA elution buffer (15mL); Protein column regeneration buffer (30mL); Protein column activation and wash buffer (60mL); Protein pH binding buffer (4mL); Protein elution buffer (8mL); Protein neutralizer (2mL); Protein loading dye (2mL); Spin columns (50); Collection tubes (200); Product inserts (1)

Rapid method for the isolation and purification of total RNA, genomic DNA and proteins sequentially from a single sample of cultured animal cells, tissue samples, blood, bacteria, yeast, fungi or plants

The total RNA, genomic DNA and proteins are all column-purified using the same column. This kit is ideal for researchers who are interested in studying the genome, proteome and transcriptome of a single biological sample, such as studies of gene expression including gene silencing experiments or mRNA knockdowns, studies involving biomarker discovery, studies in epigenetics and for characterization of cultured cell lines.

  • Fast and easy processing using rapid spin-column format
  • All columns for RNA, DNA and protein purification provided
  • Sequential isolation of nucleic acids and proteins from a single lysate, no need to split the lysate
  • Isolate all sizes of RNA, including large mRNA and rRNA to microRNA (miRNA)
  • Isolate high quality total RNA
  • Isolate high quality genomic DNA with a molecular weight ≥ 30kb
  • High yields of isolated proteins
  • No phenol or chloroform extractions

Applications:

Purifed RNA, DNA and protein suitable for assays such as PCR, real-time PCR, reverse transcription PCR, Northern blotting, RNase protection, expression array assays, DNA sequencing, Southern blotting, SNP analysis, SDS-PAGE and Western Blots

TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
For Use With (Application) PCR, RT-PCR, reverse transcription PCR, northern blotting, RNase protection, expression array assays, DNA sequencing, southern blotting, SNP analysis, SDS-PAGE and western Blots
Includes Lysis solution 40mL, RNA wash solution 20mL, RNA elution solution 6mL, genomic DNA wash solution 15mL, genomic DNA Elution Buffer 15mL, Protein column regeneration Buffer 30mL, Protein column activation and Wash Buffer 60mL, Protein pH Binding Buffer 4mL, Protein Elution Buffer 8mL, Protein neutralizer 2mL, Protein loading dye 2mL, spin columns 50, collection tubes 200, product inserts 1
No. of Reactions 50 Preps
Quantity 50 Preps
Yield 15μg RNA HeLa cells (1 x 106 cells), 150μg Protein HeLa cells (1 x 106 cells)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.