Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher BioReagents™ SurePrep™ RNA/Protein Purification Kit

Catalog No. BP280650 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 Preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP280650 50 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BP280650

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Lysis solution (40mL); Nucleic acid wash solution (22mL); Nucleic acid elution buffer (15mL); Protein column regeneration buffer (30mL); Protein column activation and wash buffer (60mL); Protein pH binding buffer (4mL); Protein elution buffer (8mL); Enzyme incubation buffer (6mL); Protein neutralizer (2mL); Protein loading dye (2mL); Spin colulmns (50); Collection tubes (150); Product insert (1)

Rapid method for the isolation and purification of total RNA and proteins sequentially from a single sample using the same column in less than 20 minutes

  • Fast and easy processing using rapid spin-column format
  • All columns for RNA purification and protein purification provided
  • Sequentially isolate high quality RNA and proteins from a single lysate – no need to split the lysate
  • Isolate all sizes of RNA, from large mRNA and rRNA down to microRNA (miRNA)
  • No phenol or chloroform extractions
  • Purify total RNA and proteins from a variety of biological sources such as cultured animal cells, tissue samples, blood, bacterica, yeast, fungi or plants

Applications:

Purified total RNA and protein from same biological sample for studies of gene expression including gene silencing experiments, mRNA knockdowns, or experiments correlating RNA and protein expression levels. RNA qualified for use in real time PCR, reverse transcription PCR, Northern blotting, RNase protection and expression array assays. Proteins suitable for use in SDS-PAGE analysis and Western blots.

TRUSTED_SUSTAINABILITY

Specifications

Format Spin Column
For Use With (Application) Purified total RNA and Protein from same biological sample for studies of gene expression including gene silencing experiments, mRNA knockdowns or experiments correlating RNA and Protein expression levels. RNA qualified for use in real time PCR, reverse transcription PCR, northern blotting, RNase protection and expression array assays. Proteins suitable for use in SDS-PAGE analysis and Western blots
Includes Lysis solution 40mL, Nucleic Acid wash solution 22mL, Nucleic Acid Elution Buffer 15mL, Protein column regeneration Buffer 30mL, Protein column activation and Wash Buffer 60mL, Protein pH Binding Buffer 4mL, Protein Elution Buffer 8mL, Enzyme incubation Buffer 6mL, Protein neutralizer 2mL, Protein loading dye 2mL, spin columns 50, collection tubes 150, product insert 1
Quantity 50 Preps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.