Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Chemical™ Gill Method Hematoxylin Stains

Catalog No. CS4014D Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Product Type:
Gill Method Hematoxylin
Gill Method Hematoxylin (Gill-2)
Gill Method Hematoxylin (Stain 1)
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
CS401-4D 4 L Gill Method Hematoxylin (Gill-2)
CS400-1D 1 L Gill Method Hematoxylin
CS400-4D 4 L Gill Method Hematoxylin
CS401-1D 1 L Gill Method Hematoxylin (Stain 1)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. CS401-4D

missing translation for 'validMainframeMsg'

Specifications

Product Type Gill Method Hematoxylin (Gill-2)
Quantity 4 L
For Use With (Application) Histological and Cytological Staining
pH 2.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.