Learn More
Fisher Container™ 1073B Tyvek Autoclavable Precision Clean Breather Bag Stoppers
Catalog No. 19351221 Shop All Fisher Container Corp. Products
Catalog No. 19-351-221 Supplier Fisher Container™ Supplier No. 91164
Specifications
Certifications/Compliance NASA
Material Tyvek, PCHD
Moisture Vapor Transmission Rate 1615g/m2/24 hr. and 0.27g/100 in.2/24 hr.
Thickness (English) 7.3/3 mil