Learn More
Bleach Solution Dispenser System
Catalog No. p-84268
Change view
Click to view available options
Product Type:
Bleach Cartridge Refills
Bleach Dilution Starter Kit
Bleach Dilution System
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-640-500 Bleach Dilution System
23-640-501 Bleach Cartridge Refills
23-640-502 Bleach Dilution Starter Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-640-500 Supplier Deardorff Fitzsimmons Corp Supplier No. 77005FS
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Eliminates waste associated with disinfecting laboratory surfaces with diluted bleach solutions
Unique, two-bottle trigger sprayer automatically mixes and dilutes 5.25% bleach with tap water to make a 10% bleach solution (5000ppm available chlorine).
  • Eliminates measuring, mixing, dating, and discarding bleach solution
  • Simply fill/refill water cartridge with tap water and lock in disposable bleach cartridge as needed
  • Sealed cartridge eliminates hazard of spilling bleach on skin and clothing
Bleach Dilution Sprayer
  • Ergonomic trigger sprayer facilitates high usage
  • Durable construction stands up to bleach
  • Includes sprayer head and water bottle
Bleach Solution Cartridge
  • 5.25% Sodium Hypochlorite Bleach
  • EPA-registered to kill Clostridium difficile spores in four minutes (EPA Reg. No. 75266-1 )
  • Each cartridge makes approximately 1 gallon of useable solution
  • Twelve 11oz. bleach cartridges
Specifications
Product Type Bleach Dilution System
Certifications/Compliance EPA
Dilution 5.25% Bleach/tap Water for 10:1 water-to-bleach solution
For Use With (Application) Eliminates waste associated with disinfecting laboratory surfaces with diluted bleach solutions
Includes Two bottle trigger sprayer head and water bottle
Test Time 30 sec. to 4 min.