Learn More
Fisher Healthcare™ Pinnacle Portfolio Eosin-Y Stain
Catalog No. 22220104 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 x 500 mL
Catalog No. Quantity
22-220-104 4 x 500 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-220-104
Add to Cart
Add to Cart
This item is not returnable. View return policy
A fast-acting, alcoholic counterstain which provides excellent delineation of cytoplasmic components
Eosin-Y Stain Features:
  • The color contrast between the nuclei and cytoplasm is unmatched
  • Various shades of red and pink differentiate muscle, red blood cells and connective tissue
  • Strict quality assurance guarantees product consistency and longevity
  • Quantitative testing eliminates the need for adjustments in dilution or time
Pinnacle Portfolio
  • Designed as a system
  • Each component has added value and benefits that reduce cost per slide
  • Unique chemical formulae of the Pinnacle Portfolio promote long-term chemical stability, which extends the usable life of each component
  • Subjected to some of the highest quality manufacturing standards in the industry
  • Packaging improvements enhance product longevity
Specifications
Product Type Eosin
Quantity 4 x 500 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov