Learn More
Fisher Healthcare™ Pinnacle Portfolio Hematoxylin 2 Stain
Catalog No. 22220102 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 x 500 mL
Catalog No. Quantity
22-220-102 4 x 500 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-220-102
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Designed to produce staining intensity similar to Harris Hematoxylin
Hematoxylin 2 Stain
  • Does not contain mercuric oxide and will not form a metallic sheen over its surface each day
  • This rapid stain is recommended for frozen sections
  • Requires no filtration prior to use
  • Will not form crystals upon standing or exposure to cold weather
  • Will stain tissue as intense as Gill 3
  • Has longer shelf life, and is more consistent and stable than Harris Hematoxylin
Pinnacle Portfolio
  • Designed as a system
  • Each component has added value and benefits that reduce cost per slide
  • Unique chemical formulae of the Pinnacle Portfolio promote long-term chemical stability, which extends the usable life of each component
  • Subjected to some of the highest quality manufacturing standards in the industry
  • Packaging improvements enhance product longevity
Differentiation is not necessary; however, Clarifier™ 2 should be used to eliminate any background staining caused by the use of excessive water bath adhesive
Specifications
Product Type Hematoxylin
Quantity 4 x 500 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov