Learn More
Fisher Healthcare™ Pinnacle Portfolio Hematoxylin Stain+
Catalog No. 22220100 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
4 x 500 mL
Catalog No. Quantity
22-220-100 4 x 500 mL
1 options
This item is not returnable. View return policy
Catalog No. 22-220-100
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Uniquely formulated product yields results which exceed that of Gill or Harris hematoxylins
Hematoxylin Stain-Plus
  • No affinity for acid mucopolysaccharide (mucin) staining
  • Slides stained with Hematoxylin+ are crisp with clear, well-defined nuclear chromatin delineation Uses aluminum ammonium sulfate at its saturation point to maintain the highest amount of dye-lake complex for optimal staining
  • Does not contain alcohol
Pinnacle Portfolio
  • Designed as a system
  • Each component has added value and benefits that reduce cost per slide
  • Unique chemical formulae of the Pinnacle Portfolio promote long-term chemical stability, which extends the usable life of each component
  • Subjected to some of the highest quality manufacturing standards in the industry
  • Packaging improvements enhance product longevity
Specifications
Product Type Hematoxylin
Quantity 4 x 500 mL
WARNING: Reproductive Harm - www.P65Warnings.ca.gov