Learn More
Fisher Healthcare™ PROTOCOL™ Define™ Histological Clarifier
Catalog No. 23310350 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
3785.4mL
Catalog No. Quantity
23-310350 3785.4mL
1 options
This item is not returnable. View return policy
Catalog No. 23-310350
This item is not returnable. View return policy
Removes residual stain that may bind to the slide adhesives
Use after specimen staining with hematoxylin. Removes residual stain that may bind to the slide adhesives. Use only with progressive staining.
Specifications
Product Type Tissue Clearing Reagent
Quantity 3785.4mL
For Use With (Application) Removes residual stain that may bind to the slide
WARNING: Reproductive Harm - www.P65Warnings.ca.gov