Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Hema 3™ Fixative and Solutions
GSA_VA

Catalog No. p-121466 Shop All Fisherbrand Products
Change view
Click to view available options
Product Type:
Hema 3 Fixative
Hema 3 Solution I
Hema 3 Solution II
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
23-122929 Hema 3 Fixative
23-122937 Hema 3 Solution I
23-122952 Hema 3 Solution II
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-122929
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For 30-second, three-step staining comparable to the Wright-Giemsa method

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 1 gal.
Boiling Point 64.7°C
Format Liquid
For Use With (Application) For manual staining
Color Clear, Light Green
Product Type Hema 3 Fixative
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.