Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Hema-G Stains for Geometric Data Hemastainer™
GSA_VA

Catalog No. 23247060 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 gal.
Product Type:
Hema-G Fixative
Hema-G Liquid Buffer
Hema-G Wright Stain, New Formulation
Hema-G Wright Stain, Regular
Hema-G Wright-Giemsa Stain, New Formulation
Hema-G Wright-Giemsa Stain, Regular
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
23-247060 1 gal. Hema-G Fixative
23-011114 1 gal. Hema-G Wright Stain, New Formulation
23-011115 1 gal. Hema-G Wright-Giemsa Stain, New Formulation
23-247057 1 gal. Hema-G Wright-Giemsa Stain, Regular
23-247056 1 gal. Hema-G Wright Stain, Regular
23-247058 1 gal. Hema-G Liquid Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 23-247060
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Quality-control tested on human blood smears to ensure proper staining and absence of precipitate

Stains for manual and automated staining methods

  • All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
  • Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
  • Carefully balanced for optimum staining pH
  • Remain stable for a minimum of 12 months when stored at room temperature
  • Allow varied staining times to suit individual preferences
  • Packaged in durable plastic bottles for safety and easy handling
  • Convenient finger tabs and clear reagent identification eliminate improper line connection

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Quantity 1 gal.
Storage Requirements Room Temperature
For Use With (Equipment) Geometric data Hemastainer™
Product Type Hema-G Fixative
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.