Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Hema-Quik™ and Hema-Quik II Stain Solutions

Catalog No. 23751784 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
90 mL
1 L
1 gal.
Product Type:
Hema-Quik II Stain Solution
Hema-Quik Stain Solution
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
23-751784 1 gal. Hema-Quik II Stain Solution
23-123745 1 L Hema-Quik Stain Solution
23-200725 1 L Hema-Quik II Stain Solution
23-123919 90 mL Hema-Quik Stain Solution
23-206441 90 mL Hema-Quik II Stain Solution
23-250466 1 gal. Hema-Quik Stain Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 23-751784
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carefully balanced for optimum staining pH

Stains for manual and automated staining methods.

  • All PROTOCOL Hematology Stains are prepared with materials certified by the Biological Stain Commission to ensure purity
  • Manufactured with strict quality control measures to ensure lot-to-lot uniformity, including extensive analytical and performance testing on human specimens
  • Remain stable for a minimum of 12 months when stored at room temperature
  • Allow varied staining times to suit individual preferences
  • Packaged in durable plastic bottles for safety and easy handling
  • Convenient finger tabs and clear reagent identification eliminate improper line connection

Specifications

Quantity 1 gal.
Storage Requirements Room Temperature
Boiling Point 65°C
Format Liquid
Color Dark Blue
Product Type Hema-Quik II Stain Solution
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.