Learn More
Fisher Healthcare™ Protocol™ HemaSpray™ Reagents for use with the Wescor™ Aerospray™ stainer
GSA_VA
Catalog No. 23316192 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 gal.
Product Type:
REAGENT A (Rinse)
REAGENT B (Blue Stain)
REAGENT C (Red Stain)
REAGENT D (Methanol)
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
23-316192 500 mL REAGENT D (Methanol)
23-033729 1 gal. REAGENT A (Rinse)
23-033730 1 gal. REAGENT B (Blue Stain)
23-033731 1 gal. REAGENT C (Red Stain)
23-033732 1 gal. REAGENT D (Methanol)
23-316189 500 mL REAGENT A (Rinse)
23-316190 500 mL REAGENT B (Blue Stain)
23-316191 500 mL REAGENT C (Red Stain)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Catalog No. 23-316192
Add to Cart
Add to Cart
This item is not returnable. View return policy
Manufactured using a two-part aqueous Romanowsky stain formulation
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Quantity 500 mL
Storage Requirements Room Temperature
Product Type REAGENT D (Methanol)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov