Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ PROTOCOL™ Saccomanno Fluid

Catalog No. 23751054 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-751054 1gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-751054

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Fixative and transport medium for cytologic specimens

Minimizes shrinkage of cells and avoids coagulation of mucoproteins.

Specifications

Boiling Point 85°C
Color Light Green
Product Type Fixative
Quantity 1gal.
For Use With (Application) Fixative and transport medium for cytologic specimens
Form Liquid
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.