Learn More
Fisher Healthcare™ Sure-Vue™ ASO Test Kit
Catalog No. 23038000 Shop All Fisherbrand Products
Change view
Click to view available options
Clia Complexity:
Moderate
Quantity:
50 Tests
Catalog No. Clia Complexity Quantity
23-038000 Moderate 50 Tests
1 options
This item is not returnable. View return policy
Catalog No. 23-038000
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
View Alternate Product
This item is not returnable. View return policy
Includes
Slides (six-well), latex reagent, positive and negative controls, and package insert
Increased sensitivity ideal for testing pediatric patients
  • CLIA Complexity: Non-Waived
  • CPT Codes: 86060/86063
  • Two-minute latex agglutination slide test
  • For the qualitative and semi-quantitative determination of antistreptolysin-O in serum
  • Can be used to screen both for levels of ASO in serum greater than 200 IU/mL and levels of ASO greater than 100 IU/mL using an alternative sampling
  • Can also be used to determine titers of ASO in serum using the semi-quantitative technique
  • For in vitro diagnostic use
Specifications
CPT Code 86060, 86063
Clia Complexity Moderate
Type Sure-Vue™ ASO Test Kit
For Use With (Application) To screen both for levels of ASO in Serum greater than 200 IU/mL and levels of ASO greater than 100 IU/mL using an alternative sampling and determine titers of ASO in Serum using the semi-quantitative technique
Includes Reagent, disposable slides and prediluted positive and negative control
Quantity 50 Tests
Test Time 2 min.