Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ H. pylori Test Kit

Catalog No. 23900535 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
30 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-535 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-535

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Test packs, developer buffer, positive and negative controls, instruction card and package insert

For qualitative detection of antibodies to H. pylori in whole blood, serum and plasma

  • CLIA Complexity: Moderate and Waived
  • CPT Codes: 86677/86318QW
  • Sensitivity: 88%†
  • Specificity: 88%†
  • Qualitative, membrane strip-based immunoassay for detection of IgG antibodies to Helicobacter pylori (H. pylori)
  • Results in 10 minutes
  • Store at room temperature or in refrigerator
  • For professional, in vitro use only

† Compared with biopsy results.

Specifications

Clia Complexity Waived, Moderate
Detectable Analytes H. pylori IgG
Type H. pylori Test Kit
Includes Test packs, developer Buffer, positive and negative control, instruction card and package insert
Quantity 30 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.