Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Mono Latex Test Kit

Catalog No. 23038006 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1000 Tests
150 Tests
50 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038006 50 Tests
23-038007 150 Tests
23-038-008 1000 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 23-038006

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

This item is not returnable. View return policy

Includes

Latex reagent, disposable slides (six-well), and prediluted positive and negative controls

Qualitative and semi-quantitative results

  • CLIA Complexity: Non-Waived
  • CPT Code: 86308
  • Latex agglutination slide test designed for the qualitative and semi-quantitative detection of infectious mononucleosis heterophile antibodies in serum and plasma
  • Test results in three minutes
  • Sensitivity of 99% and specificity of 99.4%
  • For in vitro diagnostic use

Specifications

Clia Complexity Non-waived
Detectable Analytes Mononucleosis Heterophil Antibodies
Type Mononucleosis Latex Test Kit
Includes Latex reagent, disposable slides (6-well) and prediluted positive and negative control
Quantity 50 Tests
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.