Learn More
Fisher Healthcare™ Sure-Vue™ RF Rapid Latex Test Kit
Catalog No. p-112682 Shop All Fisherbrand Products
Change view
Click to view available options
Clia Complexity:
Moderate
Quantity:
100 Tests
Type:
RF Rapid Latex Test Kit
Catalog No. Clia Complexity Quantity Type
23-038002 Moderate 100 Tests RF Rapid Latex Test Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-038002
This item is not returnable. View return policy
Includes
Test kits include latex reagent, disposable slides (six-well), prediluted positive and negative controls, and package insert
Suspension of latex particles is coated with human immunoglobulin
  • CLIA Complexity: Non-Waived
  • CPT Codes: 86430/86318
  • Latex agglutination test for the qualitative and semi-quantitative determination of rheumatoid factor in serum
  • Can be used to both screen for levels of RF in serum greater than 10 IU/mL and determine titers of RF in serum in IU/mL using the semi-quantitative technique
  • For in vitro diagnostic use
Specifications
CPT Code 86430, 86318
Clia Complexity Moderate
Detectable Analytes Rheumatoid Factor (RF)
Type RF Rapid Latex Test Kit
Includes Latex reagent, disposable slides (six-well), prediluted positive and negative control
Quantity 100 Tests
Test Time 2 min.