Learn More
Fisher Healthcare™ Sure-Vue™ Rota Test Kit
Catalog No. SA069111 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 Tests
Catalog No. Quantity
SA069111 25 Tests
1 options
This item is not returnable. View return policy
Catalog No. SA069111
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
20 test devices, tubes, extraction buffer, disposable sample transfer pipets, collection swabs, and package insert
Simple two-step procedure.
  • CLIA Complexity: Non-Waived
  • CPT Code: 87425
  • Rapid membrane-based immunogold assay for the qualitative detection of rotavirus antigen in human fecal specimens; aids in the diagnosis of acute gastroenteritis caused by rotavirus infection
  • Preliminary test results in 30 seconds; final results in 15 minutes
  • Sensitivity and specificity of 100% compared to electronmicroscopy
  • For in vitro diagnostic use only
Specifications
CPT Code 87425
Clia Complexity Non-waived
Detectable Analytes Rotavirus Antigen
Type Rotavirus Test Kit
Includes 20 Each test devices, tubes, extraction Buffer, disposable sample transfer pipets, collection swabs and one package insert
Quantity 25 Tests
Test Time 15 min.