Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ RPR Disposable Slides, 50 Pk.
SDP

Catalog No. 23038013 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 Pack
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038013 50 Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-038013
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

  • Antigen detects “reagin” present in serum of syphilitic patients
  • For in vitro diagnostic use

Specifications

Description Disposable Slide
Detectable Analytes Reagin (Syphilis)
Type Disposable Slide
For Use With (Application) For in-vitro diagnostic use
Includes Disposable slides
Quantity 50 Pack
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.