Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ RPR Control Set
GSA_VASDP

Catalog No. 23038012 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 x 1.5 mL
3 x 1.5 mL
Description:
RPR Control
RPR Titer Control
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Description
23-038012 3 x 1.5 mL RPR Control
23-029-966 1 x 1.5 mL RPR Titer Control
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-038012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Variation of the standard VDRL

  • CLIA Complexity: Non-Waived
  • CPT Codes: 86542/86593
  • Cardiolipin charcoal microparticle test for the qualitative and semi-quantitative determination of reagin, which is associated with syphilis
  • Antigen detects 'reagin' present in serum of syphilitic patients
  • Test results in eight minutes
  • Positive results yield a flocculation of the antigen; negative results yield no reaction and a gray homogeneous suspension, or a button of non-aggregating carbon particles in the center of the circle
  • For in vitro diagnostic use

Contains Sodium Azide 0.1%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Description RPR Control
Detectable Analytes Reagin (Syphilis)
Type RPR Control
For Use With (Application) For in-vitro diagnostic use
Quantity 3 x 1.5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.