Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Rubella Latex Test Kit

Catalog No. 23038004 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038004 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-038004

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Latex reagent, disposable slides (15-well), spreading pipets, diluent buffer, high and low positive controls, negative control, and package insert

Requires a dilution step for 10 IU sensitivity

  • CLIA Complexity: Non-Waived
  • CPT Codes: 86762/86406
  • Sensitivity: 99.5%
  • Specificity: 96.1%
  • Non-dilution: 1-2 IU sensitivity; dilution: 10 IU sensitivity
  • Latex agglutination slide test for the qualitative and semi-quantitative determination of rubella virus antibodies in serum
  • Test results in eight minutes
  • Reagent is a suspension of polystyrene latex particles of uniform size coated with soluble rubella virus antigen from disrupted virus
  • For in vitro diagnostic use

Specifications

CPT Code 86762, 86406
Clia Complexity Non-waived
Detectable Analytes Rubella Virus Antibodies
Type Rubella Latex Test Kit
Includes Latex reagent, disposable slides (15-well), spreading pipets, diluent Buffer, high and low positive control and negative control
Quantity 100 Tests
Test Time 8 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.