Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Sure-Vue™ Signature Rubella Latex Test

Catalog No. 23038060 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-038-060 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-038-060

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Latex reagent, high positive control, low positive control, negative control, diluent, disposable test cards, and package insert

Does not require a dilution step for 10 IU sensitivity

  • CLIA Complexity: Non-Waived
  • CPT Codes: 86762/86406
  • Sensitivity: 100%
  • Specificity: 98.6%
  • Rapid latex agglutination test for the qualitative and semi-quantitative detection of both IgG and IgM anti-rubella virus antibodies in serum
  • Results in five minutes
  • For in vitro diagnostic use

Specifications

CPT Code 86762, 86406
Clia Complexity Non-waived
Detectable Analytes Rubella IgG, Rubella IgM
Type Rubella Latex Test Kit
Includes Latex reagent, high positive control, low positive control, negative control, diluent, disposable test cards and package insert
Quantity 100 Tests
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.