Learn More
Fisher Healthcare™ Sure-Vue™ Serum/Urine hCG Test Kit
Catalog No. 23900529 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
30 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-529 50 Tests
23-900-528 30 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-529
Add to Cart
Add to Cart
This item is not returnable. View return policy
One-step test for qualitative detection of hCG in serum or urine to aid in early detection of pregnancy
  • CLIA Complexity: Waived (Urine); Moderate (Serum)
  • Sensitivity of 25mIU/mL
  • Results in three minutes for urine or five minutes for serum
  • Combination of monoclonal and polyclonal antibody reagent selectively detects elevated levels of hCG
  • Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line at the T (test) area
  • Absence of this colored line suggests a negative result
  • A colored line at the C (control) area will always appear regardless of the presence or absence of hCG
  • Urine specimens collected at any time of day can be used
  • For professional in vitro diagnostic use only
  • Addition of LH (300mIU/mL), FSH (1000mIU/mL) and TSH (1000μIU/mL) to negative or positive urine/serum showed no cross-reactivity with hCG
  • Serum should be collected without anticoagulants and hemolysis should be avoided
Specifications
Detectable Analytes hCG
Sample Type Serum, Urine
Type Serum and Urine hCG Test Kit
Quantity 50 Tests
Test Time 3 min. for urine, 5 min. for serum