Description
- CLIA Complexity: Waived (Urine); Moderate (Serum)
- Sensitivity of 25mIU/mL
- Results in three minutes for urine or five minutes for serum
- Combination of monoclonal and polyclonal antibody reagent selectively detects elevated levels of hCG
- Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line at the T (test) area
- Absence of this colored line suggests a negative result
- A colored line at the C (control) area will always appear regardless of the presence or absence of hCG
- Urine specimens collected at any time of day can be used
- For professional in vitro diagnostic use only
- Addition of LH (300mIU/mL), FSH (1000mIU/mL) and TSH (1000μIU/mL) to negative or positive urine/serum showed no cross-reactivity with hCG
- Serum should be collected without anticoagulants and hemolysis should be avoided
Specifications
Specifications
| Detectable Analytes | hCG |
| Sample Type | Serum, Urine |
| Type | Serum and Urine hCG Test Kit |
| Quantity | 30 Tests |
| Test Time | 3 min. for urine, 5 min. for serum |