Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Signature Strep A Test Kit
GSA_VA

Catalog No. 23200276 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-200-276 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-200-276
  • $626.00
    $384.65 / Pack of 50

    Save $241.35

    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $8,657.00
    $6,923.70 / Case of 18 PK

    Save $1,733.30

    20% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Test sticks, reagents, controls, workstation, package insert, swabs and test tubes

Rapid, lateral flow test with 100% specificity. Sure-Vue™ Signature Strep A Test Kit is a color immunochromatographic assay intended for the qualitative detection of Group A Streptococcus antigen directly from throat swab specimens.

  • CLIA Complexity: Waived
  • CPT Code: 87880QW
  • Sensitivity: 95.7%
  • Specificity: 100.0%
  • Test Time: five minutes
  • Sample Type: throat swabs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Accuracy 95.7% (Not statistically different than single swab culture)
Content And Storage Room Temperature (15°C to 30°C, 59°F to 86°F)
CPT Code 87880QW
Clia Complexity Waived
Detectable Analytes Streptococci (Group A)
Format Immunochromatographic Dipstick
Sample Type Throat Swab
Sensitivity 95.7%
Type Streptococcus A Test Kit
Includes Test sticks, reagents, control, workstation, package insert, swabs and test tubes
Packaging Type Test Kit
Quantity 50 Tests
Specificity 100%
Test Time 5 min.
Analysis Time 5 min.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.