Learn More
Fisher Healthcare™ Sure-Vue™ STAT Serum/Urine hCG Test Kit
Catalog No. 23900530 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
30 Tests
50 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-530 30 Tests
23-900-531 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-530
Add to Cart
Add to Cart
This item is not returnable. View return policy
Detects pregnancy as early as first day of missed menses
  • CLIA Complexity: Waived (Urine); Moderate (Serum)
  • Sensitivity of 20mIU/mL for urine and 10mIU/mL for serum
  • Results in three minutes for urine or five minutes for serum
  • Visual, one-step test for the qualitative determination of human chorionic gonadotropin (hCG) in serum or urine to aid in early detection of pregnancy
  • Combination of monoclonal and polyclonal antibody reagent selectively detects elevated levels of hCG
  • Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line at the T (test) area
  • Absence of this colored line suggests a negative result
  • A colored line at the C (control) area will always appear regardless of the presence or absence of hCG
  • Urine specimens collected at any time of day can be used
  • Addition of LH (300mIU/mL), FSH (1000mIU/mL) and TSH (1000μIU/mL) to negative or positive urine/serum showed no cross-reactivity with hCG
  • Serum should be collected without anticoagulants and hemolysis should be avoided
  • For professional in vitro diagnostic use only
Specifications
Detectable Analytes hCG
Sample Type Serum, Urine
Type Serum and Urine Test Kit
Quantity 30 Tests
Test Time 3 min. for urine, 5 min. for serum