Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Strep A Lateral Flow Test Kit

Catalog No. 23900532 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
27 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-900-532 27 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-900-532
  • $258.00
    $183.65 / Pack of 27

    Save $74.35

    29% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
 Restricted Item
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

27 test packs and throat swabs, extraction reagent, package insert, instruction card and positive and negative controls

One-step lateral-flow immunoassay for the qualitative detection of Group A Strep from throat swabs

  • CLIA Complexity: Waived
  • CPT Code: 87081
  • Sensitivity: 97%
  • Specificity: 95%
  • Intended for use as an aid in the diagnosis of Group A Streptococcal infection
  • Results in five minutes
  • Membrane strip coated with rabbit anti-Strep A antibody on the test band region and goat anti-rabbit antibody on the control band region
  • Control band verifies sufficient volume has been added and proper flow is obtained
  • For in vitro diagnostic use only

Specifications

CPT Code 87081
Clia Complexity Waived
Detectable Analytes Streptococci (Group A)
Sample Type Throat Swab
Type Streptococcus A Lateral Flow Test Kit
Includes 27 Test packs and throat swabs, extraction reagent, package insert, instruction card and positive and negative control
Quantity 27 Tests
Test Time 5 min.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.