Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Healthcare™ Sure-Vue™ Serum Test Kit Control
GSA_VA

Catalog No. sa087712 Shop All Fisherbrand Products
Change view
Click to view available options
Type:
STAT hCG Control Test Kit
Serum hCG Control Test Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
SA087712 Serum hCG Control Test Kit
SA087715 STAT hCG Control Test Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SA087712
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Includes

Negative, Low Positive and High Positive levels; 3 x 5mL

Specifically for use with Sure-Vue hCG Test Kits

For professional in vitro diagnostic use only

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

For Use With (Equipment) Sure-Vue™ Serum hCG test kit
Sample Type Serum
Type Serum hCG Control Test Kit
Includes Negative, low positive and high positive control
Quantity 3 x 5 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.