Learn More
Fisher Healthcare™ Sure-Vue™ Urine hCG Test Kits
Catalog No. 23900526 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 Tests
30 Tests
Type:
Urine hCG Dipstick Test
Urine hCG Test Kit
Description:
Urine hCG dipstick test
Urine hCG test kit
This item is not returnable. View return policy
This item is not returnable. View return policy
Includes
Test devices (cassettes or dipsticks), disposable specimen droppers and package insert
Greater than 99% accuracy
  • CLIA Complexity: Waived
  • Results in three minutes
  • Visual, one-step test for the qualitative determination of human chorionic gonadotropin (hCG) in urine to aid in early detection of pregnancy
  • Utilizes combination of monoclonal and polyclonal antibody reagent to selectively detect elevated levels of hCG
  • Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line at the T (test) area
  • Absence of this colored line suggests a negative result
  • A colored line at the C (control) area will always appear, regardless of the presence or absence of hCG
  • Sensitivity of 25mIU/mL
  • Addition of LH (300mIU/mL), FSH (1000mIU/mL) and TSH (1000μIU/mL) to negative or positive urine showed no cross-reactivity with hCG
  • For professional in vitro diagnostic use only
Specifications
Accuracy >99%
Description Urine hCG test kit
Detectable Analytes hCG
Sample Type Urine
Type Urine hCG Test Kit
Includes Test device (cassettes or dipsticks), disposable specimen droppers
Quantity 30 Tests
Test Time 3 min.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.