Learn More
Fisher Healthcare™ Sure-Vue™ Urine hCG Test Kits
Catalog No. 23900527 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 Tests
30 Tests
Type:
Urine hCG Dipstick Test
Urine hCG Test Kit
Description:
Urine hCG dipstick test
Urine hCG test kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Type Description
23-900-527 25 Tests Urine hCG Dipstick Test Urine hCG dipstick test
23-900-526 30 Tests Urine hCG Test Kit Urine hCG test kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-900-527
Add to Cart
Add to Cart
This item is not returnable. View return policy
Includes
Test devices (cassettes or dipsticks), disposable specimen droppers and package insert
Greater than 99% accuracy
  • CLIA Complexity: Waived
  • Results in three minutes
  • Visual, one-step test for the qualitative determination of human chorionic gonadotropin (hCG) in urine to aid in early detection of pregnancy
  • Utilizes combination of monoclonal and polyclonal antibody reagent to selectively detect elevated levels of hCG
  • Positive specimens react with the specific antibody-hCG colored conjugate and form a colored line at the T (test) area
  • Absence of this colored line suggests a negative result
  • A colored line at the C (control) area will always appear, regardless of the presence or absence of hCG
  • Sensitivity of 25mIU/mL
  • Addition of LH (300mIU/mL), FSH (1000mIU/mL) and TSH (1000μIU/mL) to negative or positive urine showed no cross-reactivity with hCG
  • For professional in vitro diagnostic use only
Specifications
Accuracy >99%
Description Urine hCG dipstick test
Detectable Analytes hCG
Sample Type Urine
Type Urine hCG Dipstick Test
Includes Test device (cassettes or dipsticks), disposable specimen droppers
Quantity 25 Tests
Test Time 3 min.