Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Bacteria Culture Kit
Shop All Fisherbrand Products

Click to view available options
:
Bacteria Culture Kit
Description
Kit includes:
- Three polystyrene Petri dishes, 100 x 15mm
- Nutrient Agar powder to create 120cc of Agar
- Plastic stir rod to aid in mixing, 6 in. (15.2cm)
- Instruction sheet for preparing Agar
Note: Cultures are not included.
Specifications
Specifications
| For Use With (Application) | Ideal for middle and high school students |
| Includes | 100 x 15mm three polystyrene petri dishes, nutrient agar powder to create 120cc of agar, 6 in. (15.2cm) plastic stir rod to aid in mixing, instruction sheet for preparing agar |
| Product Type | Bacteria Culture Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction