Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ ChemAlert™ Tape
GREENER_CHOICESDPSCI_ED

Catalog No. s68160 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Blue, Red, Yellow, White, Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
S68160 Blue, Red, Yellow, White, Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S68160
Only null left
Add to Cart
Add to Cart

Color-coded, pressure-sensitive tape set.

  • Color-coded tape set includes one 14 yd. (13 m) roll each of blue, red, yellow and white; and two rolls of gray.

Specifications

Color Blue, Red, Yellow, White, Gray
Format Tape
Product Type Tape
Sterility Non-sterile
For Use With (Application) Chemical Hazard
Includes 14 yd. Roll each of blue, red, yellow and white; and two rolls of gray
Quantity 6/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.