Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Environmental Engineering Oil Spill Management Kit
SCI_ED

Catalog No. s02643 Shop All Fisherbrand Products
Change view
Click to view available options
Grade:
7 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02643 7 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Choking hazard (1) Small parts. Not for children under 3 yrs.

Catalog No. S02643
Only null left
Add to Cart
Add to Cart

Choking hazard (1) Small parts. Not for children under 3 yrs.

Includes

  • Instructor manual with reproducible student data sheets
  • Test Tubes/Stoppers
  • Vegetable Oil
  • Stopwatch
  • Test Tube Rack
  • Graduated Cylinders
  • Corn Syrup
  • Food Coloring
  • Empty Bottles with caps
  • Potting Soil
  • Aluminum Pan
  • Sand
  • Gravel
  • Pipettes
  • Spoons
  • Paper Towel Strips
  • Tubing
  • Fabric (Cotton, Polyester)
  • Deli Container
  • Thermometer
  • Sugar Packet
  • Digital Scale, 500g
  • Yeast Packet
  • Strainer
  • Dishwashing Liquid
  • Cocoa Powder
  • Stainless Steel Scoop
  • Gloves
  • Nylon Stocking
  • Cotton Balls
  • Beaker
  • Shredded Copy Paper
  • Scissors
  • Weigh Dish
  • Mixed Seashells

Hands-on oil spill awareness and cleanup kit designed for grades 7 to 12.

Students simulate deep-ocean and beach crude oil spills as they engage in the work of an environmental engineering team using models to learn the physical and chemical properties of oil and experience how scientists respond to oil spill disasters.

  • Teaches environmental impact, basic oil biology, micelle structures and chemistry concepts such as hydrophobic and hydrophilic interactions
  • Students simulate basic oil containment and removal techniques using their own oil spill models
  • Includes a quantitative analysis of absorbent materials, hands on skimming and dispersant activities and an experimental introduction to concept of bioremediation
  • Students work as team to research and systematically design oil spill solution, use iterative cycle to improve their solutions and learn to argue their points of view
  • Great for applying knowledge and teaching higher order thinking skills including creative thinking, reasoning, decision making and problem solving
  • Plan to spend several weeks on unit
  • Class size: 24 students

Order Info

Designed to accommodate 24 students working in groups of two.

Specifications

Grade 7 to 12
Product Type Kit, Oil Spill Management
Includes Quantitative analysis of absorbent materials, hands-on skimming and dispersant activities and an experimental introduction to the concept of bioremediation
For Use With (Application) Students will simulate deep ocean and beach crude oil spills
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.