Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Forensics Using Simulated Blood Kit
GREENER_CHOICESCI_ED

Catalog No. S96794 Shop All Aldon Products
Change view
Click to view available options
Grade:
7 to 14
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S96794 7 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Phenolphthalein and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm.
Catalog No. S96794 Supplier Aldon Corporation Supplier No. IS3104
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

WARNING: This product can expose you to chemicals including Phenolphthalein and Methanol/ Methyl Isobutyl Ketone, which are known to the State of California to cause cancer, birth defects and reproductive harm.

Enjoy hands-on activities and learn the use of blood type in a forensic investigation.

  • First, a presumptive blood test is used to determine if a substance on a stained piece of cloth may be blood
  • Then, they determine the blood type of the sample collected at the scene
  • Results are compared to those of samples provided from two suspects
  • Students act as lab technicians and assist investigators by examining evidence collected at a crime scene
  • Kit provides the most accurate simulation of the actual blood typing procedure
  • No toothpicks, stirring or waiting for results
  • Contains materials for 10 groups
  • Uses simulated blood; These activities do not work with real blood
  • Kit Includes:
    • 70% Ethanol (1 Bottle)
    • Blood detection reagent No. 1 (1 Bottle)
    • Blood detection reagent No. 2 (1 Bottle)
    • Simulated anti-A serum (1 Bottle)
    • Simulated anti-B serum (1 Bottle)
    • Simulated anti-Rh serum (1 Bottle)
    • Simulated Blood Samples: victim, suspect No. 1, suspect No. 2, and crime scene evidence
    • 40 blood typing trays
    • 10 cotton swabs
    • Cloth piece
    • Teacher's manual and student study guide copy-masters

Specifications

Certifications/Compliance 49 CFR 173.4
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Accurate simulation of the blood typing procedure, understanding the use of blood type in a forensic investigation
Quantity 1 Kit
Dimensions (L x W x H) 9.75 x 6.25 x 4 in.
Class Size 10 Lab Groups
Product Type Forensics Using Simulated Blood Typing Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.